Assay Details
| Gene Symbol: MSRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034810.2 | NP_001029982.2 | 1 - 2 | 210 | 72 | |
| GenBank mRNA | BC105188.1 | - | 1 - 2 | 158 | 72 | |
| CK836798.1 | - | 1 - 2 | 210 | 72 | ||
More Information
Set Membership: |
|

MSRB1
Genomic Map