Assay Details
| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035012.1 | NP_001030184.1 | 7 - 8 | 1052 | 65 | |
| GenBank mRNA | BC102809.1 | - | 7 - 8 | 1052 | 65 | |
More Information
Set Membership: |
|

RIPK1
Genomic Map