Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035013.2 | NP_001030185.1 | 6 - 7 | 952 | 97 | |
| GenBank mRNA | BC105224.1 | - | 6 - 7 | 953 | 97 | |
More Information
Set Membership: |
|

MTRF1
Genomic Map