Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035013.2 | NP_001030185.1 | 9 - 10 | 1297 | 71 | |
| GenBank mRNA | BC105224.1 | - | 9 - 10 | 1298 | 71 | |
More Information
Set Membership: |
|

MTRF1
Genomic Map