Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035033.2 | NP_001030205.1 | 8 - 9 | 1439 | 67 | |
| GenBank mRNA | BC102774.1 | - | 8 - 9 | 1440 | 67 | |
More Information
Set Membership: |
|

OMA1
Genomic Map