Assay Details
| Gene Symbol: TCP11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205472.1 | NP_001192401.1 | 6 - 7 | 965 | 63 | |
| GenBank mRNA | BC102867.1 | - | 6 - 7 | 957 | 63 | |
More Information
Set Membership: |
|

TCP11L1
Genomic Map