Assay Details
| Gene Symbol: ANP32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035074.1 | NP_001030246.1 | 1 - 2 | 290 | 67 | |
| GenBank mRNA | BC102954.1 | - | 1 - 2 | 290 | 67 | |
More Information
Set Membership: |
|

ANP32B
Genomic Map