Assay Details
| Gene Symbol: PSMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035083.1 | NP_001030255.1 | 2 - 3 | 176 | 59 | |
| GenBank mRNA | BC102595.1 | - | 2 - 3 | 176 | 59 | |
More Information
Set Membership: |
|

PSMC4
Genomic Map