Assay Details
| Gene Symbol: DARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035085.1 | NP_001030257.1 | 5 - 6 | 464 | 106 | |
| GenBank mRNA | BC103319.1 | - | 5 - 6 | 464 | 106 | |
More Information
Set Membership: |
|

DARS1
Genomic Map