Assay Details
| Gene Symbol: CIAO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035110.1 | NP_001030282.1 | 2 - 3 | 320 | 120 | |
| GenBank mRNA | BC102256.1 | - | 2 - 3 | 320 | 120 | |
| BC151428.1 | - | 2 - 3 | 317 | 120 | ||
More Information
Set Membership: |
|

CIAO2A
Genomic Map