Assay Details
| Gene Symbol: PSMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035111.3 | NP_001030283.2 | 5 - 6 | 867 | 63 | |
| GenBank mRNA | BC103400.1 | - | 5 - 6 | 836 | 63 | |
More Information
Set Membership: |
|

PSMD8
Genomic Map