Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035280.1 | NP_001030357.1 | 2 - 3 | 246 | 145 | |
| GenBank mRNA | BC102276.1 | - | 2 - 3 | 246 | 145 | |
More Information
Set Membership: |
|

RTRAF
Genomic Map