Assay Details
| Gene Symbol: EDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035307.2 | NP_001030384.1 | 1 - 2 | 172 | 60 | |
| GenBank mRNA | BC102246.1 | - | 1 - 2 | 174 | 60 | |
More Information
Set Membership: |
|

EDF1
Genomic Map