Assay Details
| Gene Symbol: HEMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035308.2 | NP_001030385.1 | 8 - 9 | 986 | 68 | |
| GenBank mRNA | BC102828.1 | - | 8 - 9 | 988 | 68 | |
More Information
Set Membership: |
|

HEMK1
Genomic Map