Assay Details
| Gene Symbol: ANXA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035325.1 | NP_001030402.1 | 5 - 6 | 388 | 64 | |
| GenBank mRNA | BC104614.1 | - | 5 - 6 | 388 | 64 | |
| JQ796276.1 | - | 3 - 4 | 311 | 64 | ||
More Information
Set Membership: |
|

ANXA3
Genomic Map