Assay Details
| Gene Symbol: PPP1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035392.2 | NP_001030469.1 | 1 - 2 | 407 | 86 | |
| GenBank mRNA | BC102669.1 | - | 1 - 2 | 408 | 86 | |
More Information
Set Membership: |
|

PPP1R2
Genomic Map