Assay Details
| Gene Symbol: LIPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035410.2 | NP_001030487.1 | 8 - 9 | 1423 | 83 | |
| GenBank mRNA | BC103072.1 | - | 8 - 9 | 1423 | 83 | |
More Information
Set Membership: |
|

LIPC
Genomic Map