Assay Details
| Gene Symbol: MTFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035412.2 | NP_001030489.1 | 5 - 6 | 634 | 54 | |
| GenBank mRNA | BC102798.1 | - | 5 - 6 | 636 | 54 | |
More Information
Set Membership: |
|

MTFR1
Genomic Map