Assay Details
| Gene Symbol: CNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035420.1 | NP_001030497.1 | 3 - 4 | 307 | 60 | |
| GenBank mRNA | BC103380.1 | - | 3 - 4 | 307 | 60 | |
More Information
Set Membership: |
|

CNN2
Genomic Map