Assay Details
| Gene Symbol: ERG28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035423.2 | NP_001030500.1 | 1 - 2 | 47 | 60 | |
| GenBank mRNA | BC102607.1 | - | 1 - 2 | 48 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102607.1, NM_001035423.2 transcripts.
Set Membership: |
|

ERG28
Genomic Map