Assay Details
| Gene Symbol: ERG28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035423.2 | NP_001030500.1 | 2 - 3 | 223 | 90 | |
| GenBank mRNA | BC102607.1 | - | 2 - 3 | 224 | 90 | |
More Information
Set Membership: |
|

ERG28
Genomic Map