Assay Details
| Gene Symbol: RIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035425.1 | NP_001030502.1 | 2 - 3 | 371 | 106 | |
| GenBank mRNA | BC107544.1 | - | 2 - 3 | 371 | 106 | |
More Information
Set Membership: |
|

RIPOR2
Genomic Map