Assay Details
| Gene Symbol: PTPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035431.2 | NP_001030508.1 | 3 - 4 | 345 | 67 | |
| GenBank mRNA | BC102127.1 | - | 3 - 4 | 346 | 67 | |
| BT030501.1 | - | 3 - 4 | 315 | 67 | ||
More Information
Set Membership: |
|

PTPN2
Genomic Map