Assay Details
| Gene Symbol: MRPS36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035438.1 | NP_001030515.1 | 2 - 3 | 109 | 71 | |
| GenBank mRNA | BC103454.1 | - | 2 - 3 | 109 | 71 | |
More Information
Set Membership: |
|

MRPS36
Genomic Map