Assay Details
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035439.1 | NP_001030516.1 | 1 - 2 | 522 | 79 | |
GenBank mRNA | BC105200.1 | - | 1 - 2 | 522 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
