Assay Details
| Gene Symbol: SH3BGRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035440.2 | NP_001030517.1 | 1 - 2 | 73 | 143 | |
| GenBank mRNA | BC103122.1 | - | 1 - 2 | 75 | 143 | |
| BT021500.1 | - | 1 - 2 | 77 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103122.1, NM_001035440.2 transcripts.
Set Membership: |
|

SH3BGRL
Genomic Map