Assay Details
| Gene Symbol: SH3BGRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035440.2 | NP_001030517.1 | 3 - 4 | 345 | 59 | |
| GenBank mRNA | BC103122.1 | - | 3 - 4 | 347 | 59 | |
| BT021500.1 | - | 3 - 4 | 349 | 59 | ||
More Information
Set Membership: |
|

SH3BGRL
Genomic Map