Assay Details
| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035442.2 | NP_001030519.1 | 3 - 4 | 344 | 102 | |
| GenBank mRNA | BC102866.1 | - | 3 - 4 | 346 | 102 | |
More Information
Set Membership: |
|

CIBAR1
Genomic Map