Assay Details
| Gene Symbol: RPP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035461.1 | NP_001030538.1 | 7 - 8 | 555 | 69 | |
| GenBank mRNA | BC103240.1 | - | 7 - 8 | 555 | 69 | |
More Information
Set Membership: |
|

RPP30
Genomic Map