Assay Details
| Gene Symbol: PRELID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035476.1 | NP_001030553.1 | 1 - 2 | 68 | 69 | |
| GenBank mRNA | BC102477.1 | - | 1 - 2 | 68 | 69 | |
| BT021687.1 | - | 1 - 2 | 126 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102477.1, NM_001035476.1 transcripts.
Set Membership: |
|

PRELID3B
Genomic Map