Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035480.2 | NP_001030557.1 | 3 - 4 | 337 | 133 | |
| GenBank mRNA | BC102676.1 | - | 3 - 4 | 338 | 133 | |
More Information
Set Membership: |
|

FAM216A
Genomic Map