Assay Details
| Gene Symbol: RPL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035503.2 | NP_001030580.1 | 3 - 4 | 240 | 79 | |
| GenBank mRNA | BC102344.1 | - | 3 - 4 | 241 | 79 | |
More Information
Set Membership: |
|

RPL28
Genomic Map