Assay Details
| Gene Symbol: RPS15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037443.2 | NP_001032520.1 | 1 | 325 | 104 | |
| GenBank mRNA | AB099015.1 | - | 1 | 313 | 104 | |
| BC108231.1 | - | 1 | 327 | 104 | ||
More Information
Set Membership: |
|

RPS15A
Genomic Map