Assay Details
| Gene Symbol: MRPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037453.2 | NP_001032530.1 | 2 - 3 | 195 | 69 | |
| GenBank mRNA | BC108174.1 | - | 2 - 3 | 196 | 69 | |
More Information
Set Membership: |
|

MRPL27
Genomic Map