Assay Details
| Gene Symbol: MON1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037454.2 | NP_001032531.1 | 3 - 4 | 549 | 156 | |
| GenBank mRNA | BC108085.1 | - | 3 - 4 | 550 | 156 | |
More Information
Set Membership: |
|

MON1B
Genomic Map