Assay Details
| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037468.1 | NP_001032545.1 | 11 - 12 | 1506 | 55 | |
| GenBank mRNA | BC108109.1 | - | 11 - 12 | 1506 | 55 | |
More Information
Set Membership: |
|

CDC7
Genomic Map