Assay Details
| Gene Symbol: FKBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037481.1 | NP_001032558.1 | 2 - 3 | 292 | 74 | |
| NM_001113256.1 | NP_001106727.1 | 3 - 4 | 395 | 74 | ||
| GenBank mRNA | BC108190.1 | - | 2 - 3 | 292 | 74 | |
| BI849286.1 | - | 3 - 4 | 381 | 74 | ||
| CN433599.1 | - | 3 - 4 | 315 | 74 | ||
| DY212243.1 | - | 3 - 4 | 469 | 74 | ||
More Information
Set Membership: |
|

FKBP2
Genomic Map