Assay Details
| Gene Symbol: OTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037484.2 | NP_001032561.1 | 1 - 2 | 153 | 83 | |
| GenBank mRNA | BC108168.1 | - | 1 - 2 | 154 | 83 | |
More Information
Set Membership: |
|

OTOR
Genomic Map