Assay Details
| Gene Symbol: CDCA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037488.2 | NP_001032565.1 | 6 - 7 | 917 | 67 | |
| GenBank mRNA | BC108118.1 | - | 6 - 7 | 918 | 67 | |
More Information
Set Membership: |
|

CDCA7
Genomic Map