Assay Details
| Gene Symbol: MRPL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037622.2 | NP_001032711.1 | 6 - 7 | 594 | 112 | |
| GenBank mRNA | BC109670.1 | - | 6 - 7 | 595 | 112 | |
More Information
Set Membership: |
|

MRPL42
Genomic Map