Assay Details
| Gene Symbol: CFI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038096.1 | NP_001033185.1 | 2 - 3 | 154 | 66 | |
| GenBank mRNA | BC108104.1 | - | 2 - 3 | 154 | 66 | |
More Information
Set Membership: |
|

CFI
Genomic Map