Assay Details
| Gene Symbol: DCTPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038202.2 | NP_001033291.1 | 2 - 3 | 251 | 136 | |
| GenBank mRNA | BC109855.1 | - | 2 - 3 | 253 | 136 | |
More Information
Set Membership: |
|

DCTPP1
Genomic Map