Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038566.1 | NP_001033655.1 | 2 - 3 | 175 | 98 | |
| GenBank mRNA | BC109636.1 | - | 2 - 3 | 175 | 98 | |
More Information
Set Membership: |
|

MED21
Genomic Map