Assay Details
| Gene Symbol: NDUFAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040488.2 | NP_001035578.1 | 4 - 5 | 510 | 116 | |
| GenBank mRNA | BC102128.1 | - | 4 - 5 | 512 | 116 | |
| X61997.1 | - | 280 | 116 | |||
More Information
Set Membership: |
|

NDUFAB1
Genomic Map