Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040502.2 | NP_001035592.1 | 1 - 2 | 151 | 66 | |
| GenBank mRNA | AJ844606.2 | - | 1 - 2 | 146 | 66 | |
| AM403243.2 | - | 1 - 2 | 148 | 66 | ||
| BC102740.1 | - | 1 - 2 | 152 | 66 | ||
| EU250279.1 | - | 1 - 2 | 130 | 66 | ||
More Information
Set Membership: |
|

ORM1
Genomic Map