Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040502.2 | NP_001035592.1 | 4 - 5 | 477 | 105 | |
| GenBank mRNA | AJ844606.2 | - | 4 - 5 | 472 | 105 | |
| AM403243.2 | - | 4 - 5 | 474 | 105 | ||
| BC102740.1 | - | 4 - 5 | 478 | 105 | ||
| EU250279.1 | - | 4 - 5 | 456 | 105 | ||
More Information
Set Membership: |
|

ORM1
Genomic Map