Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040515.2 | NP_001035605.1 | 2 - 3 | 358 | 109 | |
| GenBank mRNA | BC102694.1 | - | 2 - 3 | 361 | 109 | |
More Information
Set Membership: |
|

TTC36
Genomic Map