Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040523.2 | NP_001035613.1 | 1 - 2 | 195 | 56 | |
| GenBank mRNA | BC109742.1 | - | 1 - 2 | 196 | 56 | |
More Information
Set Membership: |
|

GLRX2
Genomic Map