Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040563.1 | NP_001035653.1 | 3 - 4 | 414 | 73 | |
| GenBank mRNA | BC104577.1 | - | 3 - 4 | 414 | 73 | |
More Information
Set Membership: |
|

HTATIP2
Genomic Map