Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040578.2 | NP_001035668.1 | 1 | 136 | 78 | |
| GenBank mRNA | BC102463.1 | - | 1 | 138 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS27L| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040578.2 | NP_001035668.1 | 1 | 136 | 78 | |
| GenBank mRNA | BC102463.1 | - | 1 | 138 | 78 | |
Set Membership: |
|